Merck
  • Determination of starane (fluroxypyr) herbicide using flow injection spectrophotometry.
Determination of starane (fluroxypyr) herbicide using flow injection spectrophotometry.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-01-25)
Jasmin Shah, M Rasul Jan, Nadia Bashir
ABSTRACT
The starane herbicide was spectrophotometrically determined by the diazotization method in a flow injection assembly. Since starane is a substituted pyridyl compound the NH2 group at the p-position was exploited for diazotization. Starane was diazotized with nitrite and the diazotized product is coupled with beta-naphthol. The absorbance of the resulting azo dye was measured at 395 nm with a molar absorptivity of 1.5 x 10(4) L mol(-1) cm(-1). The calibration graph was linear over the range of 0.6 to 10 microg/mL, with a relative standard deviation (RSD) of 1.67% and a sampling through put of 60 samples h(-1). The % recovery for the determination of starane was found to be 96%. The method was successfully applied to the determination of the active ingredient of starane herbicide in its formulation as well as in food samples.
Product Description
Supelco
Fluroxypyr, PESTANAL®, analytical standard