- Hepatotoxic cyanobacterial blooms in the lakes of northern Poland.
Hepatotoxic cyanobacterial blooms in the lakes of northern Poland.
The lakes of northern Poland are among the recreational sites most valued by Polish and German holiday makers. Given the socioeconomic importance of these lakes, water quality should be maintained at high levels for such intensive recreational purposes. In 2002 studies of species composition, biomass, and toxin production by phytoplankton and the attendant physicochemical variables were performed in order to assess the risk of cyanobacterial blooms in selected northern lakes: Lakes Jeziorak, Jagodne, Szymoneckie, Szymon, Taltowisko, Siecino, and Trzesiecko. The research showed that total phosphorus (0.1 mg P/L) and total nitrogen (1.5 mg N/L) in the studied lakes almost exceeded the permissible limits for eutrophication of water bodies. Most phytoplankton samples were taken in late summer, when cyanobacteria were expected to reach their highest biomass. At the time of sampling most of the lakes were dominated by oscillatorialean and nostocalean species. Average chlorophyll-a concentration was higher than 10 microg/L in almost all the lakes studied, which corresponded with an average microcystin concentration in the range of 4-5 microg/L. The main microcystins in the analyzed samples were dmMC-RR, MC-RR, MC-YR, and MC-LR. The results demonstrated a potential for intensive cyanobacterial blooms to appear during the summer in northern Polish lakes. The levels of cyanobacteria found in the lakes investigated indicated that toxicity had reached the first-alert level according to World Health Organization recommendations. If microcystin-producing cyanobacteria dominate, with a microcystin concentration of 2-4 microg/L, symptoms of toxicity can appear in the swimmers most sensitive to exposure. Analysis of cyanobacterial assemblages in northern Polish lakes also indicated a significant presence of Aphanizomenon species including a Scandinavian species, A. skujae (Skuja) Kom.-Legn. & Cronb. Future investigations of Polish lakes also should assess neurotoxins and study the biology of their producers. This study was the first attempt to evaluate the potential danger of toxic cyanobacterial blooms in the lakes of northern Poland.