Merck
  • The residue levels of narasin in eggs of laying hens fed with unmedicated and medicated feed.
The residue levels of narasin in eggs of laying hens fed with unmedicated and medicated feed.
Molecular nutrition & food research (2004-11-13)
Mervi Rokka, Susanna Eerola, Ulla Perttilä, Laila Rossow, Eija Venäläinen, Eija Valkonen, Jarmo Valaja, Kimmo Peltonen
ABSTRACT
Laying hens were fed contaminated feed containing narasin 2.5 mg/kg for 21 days followed by a 7 day withdrawal period, hens in the control group were fed unmedicated feed. Eggs were collected during trial days 0, 3, 7, 14, 21 and after the withdrawal period of 7 days. The concentration of narasin in yolks and egg whites was analyzed by a liquid chromatography-mass spectrometry method. Narasin was found to accumulate in yolks, where the narasin concentration increased during the treatment. The concentration of narasin varied from 5.9 to 13.8 microg/kg (mean 10.6 microg/kg) in yolks after 21 day feeding periods. The concentrations of narasin ranged from < 0.9 to 1.4 microg/kg after the withdrawal period. Narasin residues were not found in egg whites of the laying hens fed contaminated feed nor in either yolks or egg whites of the laying hens fed unmedicated feed. The effect of cooking was also tested on the amount of narasin residues in eggs. Cooking for 10 min did not significantly influence the narasin residues in eggs. Traces of lasalocid were also found in the yolks. The traces of lasalocid are attributable to an accidental contamination of the feed during its manufacture.
Sigma-Aldrich
Narasin from Streptomyces auriofaciens, ≥98% (HPLC)