Merck
  • Efflux properties of basolateral peptide transporter in human intestinal cell line Caco-2.
Efflux properties of basolateral peptide transporter in human intestinal cell line Caco-2.
Pflugers Archiv : European journal of physiology (2004-09-02)
Megumi Irie, Tomohiro Terada, Masahiro Okuda, Ken-Ichi Inui
ABSTRACT
Small peptides and some pharmacologically active compounds are absorbed from the small intestine by the apical H(+)-coupled peptide transporter 1 (PEPT1) and the basolateral peptide transporter. Here we investigated the efflux properties of the basolateral peptide transporter in Caco-2 cells using two strategies, efflux measurements and a kinetic analysis of transepithelial transport of glycylsarcosine (Gly-Sar). [(14)C]Gly-Sar efflux through the basolateral membrane was not affected significantly by the external pH. Both approaches revealed that the basolateral peptide transporter was saturable in the efflux direction, and that the affinity was lower than that in the influx direction. For two peptide-like drugs, there was no difference in substrate recognition by the basolateral peptide transporter between the two sides of the membrane. Using the kinetic parameters of PEPT1 and the basolateral peptide transporter, a computational model of Gly-Sar transport in Caco-2 cells was constructed. The simulation fitted the experimental data well. Our findings suggested that substrate affinity of the basolateral peptide transporter is apparently asymmetric, but pH-dependence and substrate specificity are symmetric for the two directions of transport. The behaviour of Gly-Sar in Caco-2 cells could be predicted by a mathematical model describing the peptide transporters.
Sigma-Aldrich
Ceftibuten hydrate, ≥98% (HPLC)