Merck
  • Haloalkane hydrolysis by Rhodococcus erythropolis cells: comparison of conventional aqueous phase dehalogenation and nonconventional gas phase dehalogenation.
Haloalkane hydrolysis by Rhodococcus erythropolis cells: comparison of conventional aqueous phase dehalogenation and nonconventional gas phase dehalogenation.
Biotechnology and bioengineering (2004-03-10)
Benjamin Erable, Isabelle Goubet, Sylvain Lamare, Marie Dominique Legoy, Thierry Maugard
ABSTRACT
Biofiltration of air polluted by volatile organic compounds is now recognized by the industrial and research communities as an effective and viable alternative to standard environmental technologies. Whereas many studies have focused on solid/liquid/gas biofilters, there have been fewer reports on waste air treatment using other biological processes, especially in a solid/gas biofilter. In this study, a comparison was made of the hydrolysis of halogenated compounds (such as 1-chlorobutane) by lyophilized Rhodococcus erythropolis cells in a novel solid/gas biofilter and in the aqueous phase. We first determined the culture conditions for the production of R. erythropolis cells with a strong dehalogenase activity. Four different media were studied and the amount of 1-chlorobutane was optimized. Next, we report the possibility to use R. erythropolis cells in a solid/gas biofilter in order to transform halogenated compounds in corresponding alcohols. The effect of experimental parameters (total flow into the biofilter, thermodynamic activity of the substrates, temperature, carbon chain length of halogenated substrates) on the activity and stability of lyophilized cells in the gas phase was determined. A critical water thermodynamic activity (a(w)) of 0.4 is necessary for the enzyme to become active and optimal dehalogenase activity for the lyophilized cells is obtained for an a(w) of 0.9. A temperature of reaction of 40 degrees C represents the best compromise between stability and activity. Activation energy of the reaction was determined and found equal to 59.5 KJ/mol. The pH effect on the dehalogenase activity of R. erythropolis cells was also studied in the gas phase and in the aqueous phase. It was observed that pH 9.0 provided the best activity in both systems. We observed that in the aqueous phase R. erythropolis cells were less sensitive to the variation in pH than R. erythropolis cells in the gas phase. Finally, the addition of volatile Lewis base (triethylamine) in the gaseous phase and the action of the lysozyme in order to permeabilize the cells was found to be highly beneficial to the effectiveness of the biofilter.
Sigma-Aldrich
1-Chlorobutane, ReagentPlus®, 99%
Sigma-Aldrich
1-Chlorobutane, biotech. grade, for protein sequence analysis, ≥99.8%
Sigma-Aldrich
1-Chlorobutane, anhydrous, 99.5%
Sigma-Aldrich
1-Chlorobutane, suitable for HPLC, ≥99.8%