Merck
  • Chain conformation of sulfated derivatives of beta-glucan from sclerotia of Pleurotus tuber-regium.
Chain conformation of sulfated derivatives of beta-glucan from sclerotia of Pleurotus tuber-regium.
Carbohydrate research (2003-12-12)
Mei Zhang, Lina Zhang, Yifeng Wang, Peter Chi Keung Cheung
ABSTRACT
Six water-insoluble (1-->3)-beta-D-glucan fractions TM8-1 to TM8-6 with weight-average molecular mass Mw ranging from 5.76 to 77.4x10(4) obtained from the sclerotia of Pleurotus tuber-regium were sulfated to produce the water-soluble fractions S-TM8-1 to S-TM8-6 with Mw from 6.0 to 64.8x10(4). The degree of substitution (DS) of S-TM8 fractions was analyzed by elemental analysis (EA) to be 1.14-1.74. The 13C NMR results indicated that the C-6 was fully substituted, and C-2, C-4 were partially substituted by the sulfo-groups. The Mw and the intrinsic viscosity [eta] of the S-TM8 fractions were measured, respectively, by size-exclusion chromatography combined with laser light scattering (SEC-LLS), LLS and viscometry in phosphate buffer solution (PBS) at 37 degrees C. The dependences of [eta] and radius of gyration <s2>z(1/2) on Mw for the S-TM8 samples were found to be [eta]=1.89x10(-2) Mw(0.70) (cm3/g) and <s2>z(1/2)=1.12x10(-4) Mw(0.81) (nm) in the Mw range tested. Based on current theories for a wormlike chain model, the molar mass per unit contour length ML and persistence length q of the S-TM8 were calculated to be 990 nm(-1) and 8.5 nm, respectively. The relatively higher q value suggested a more expanded flexible chain of S-TM8 in PBS. The water-solubility and relatively expanded chain conformation of the STM8 fractions were considered to be significant to their antiviral activity.
Sigma-Aldrich
Chlorosulfonic acid, 99%