Merck
  • Preferential block of T-type calcium channels by neuroleptics in neural crest-derived rat and human C cell lines.
Preferential block of T-type calcium channels by neuroleptics in neural crest-derived rat and human C cell lines.
Molecular pharmacology (1992-08-01)
J J Enyeart, B A Biagi, B Mlinar
ABSTRACT
We have used the whole-cell version of the patch-clamp technique to analyze the inhibition of Ca2+ currents by antipsychotic agents in neural crest-derived rat and human thyroid C cell lines. Diphenylbutylpiperidine (DPBP) antipsychotics, including penfluridol and fluspirilene, potently and preferentially block T-type Ca2+ current in the rat medullary thyroid carcinoma 6-23 (clone 6) cell line. When step depolarizations were applied at 0.1 Hz from a holding potential of -80 mV, with 10 mM Ca2+ as the charge carrier, the DPBP penfluridol inhibited T-type current with an IC50 of 224 nM. High voltage-activated L and N currents were less potently blocked. At a concentration of 500 nM, penfluridol inhibited 78.0 +/- 2.3% (n = 29) of inactivating T-type Ca2+ current, whereas the sustained high voltage-activated current was reduced by 25.6 +/- 3.5% (n = 28). Block of T-type current by penfluridol was enhanced by depolarizing test pulses applied at frequencies above 0.03 Hz. The use-dependent component of block was largely reversed by pulse-free periods at -80 mV. T-type Ca2+ channels in the human TT C cell line were blocked by penfluridol, and the potency was enhanced by reduction of extracellular Ca2+. Non-DPBP antipsychotics, including haloperidol, clozapine, and thioridazine, also blocked T-type channels, but these were 20-100 times less potent than the DPBPs. These results identify the DPBPs as a new class of organic Ca2+ channel antagonists, which are distinctive in their ability to preferentially block T-type channels. These agents will be useful in defining the function of T channels in various excitable cells. Their potent block of T-type Ca2+ channels, which would be enhanced in rapidly firing cells, suggests that this action may be relevant to the therapeutic or toxic effects of these drugs when used in clinical pharmacology.
Sigma-Aldrich
Penfluridol, ≥97% (HPLC), powder