Merck
  • Formation of an unusual dimeric compound by lead tetraacetate oxidation of a corynanthe-type indole alkaloid, mitragynine.
Formation of an unusual dimeric compound by lead tetraacetate oxidation of a corynanthe-type indole alkaloid, mitragynine.
Chemical & pharmaceutical bulletin (2002-07-20)
Hiromitsu Takayama, Hayato Ishikawa, Mariko Kitajima, Norio Aimi
ABSTRACT
Lead tetraacetate oxidation of a Corynanthe-type indole alkaloid, mitragynine, produced mainly 7-acetoxyindolenine derivative (2) together with a dimeric compound (4) as a minor product. The novel structure having a bridge between the C-11' and C-7 positions in the respective indolenine parts and its formation mechanism were studied.
Sigma-Aldrich
1,3-Dimethoxybenzene, ≥98%, FG
Sigma-Aldrich
1,3-Dimethoxybenzene, ≥98%