Merck
  • Effect of methacrylonitrile on cytochrome P-450 2E1 (CYP2E1) expression in male F344 rats.
Effect of methacrylonitrile on cytochrome P-450 2E1 (CYP2E1) expression in male F344 rats.
Journal of toxicology and environmental health. Part A (2002-04-10)
Hongbing Wang, Brian Chanas, Burhan I Ghanayem
ABSTRACT
Tissue-specific induction of cytochrome P-450s (CYP) followed by increased in situ bioactivation may contribute to chemical-induced site-specific toxicity. In rats, methacrylonitrile (MAN) is metabolized by cytochrome P-450 2E1 (CYP2E1) to acetone, which is eliminated along with parent MAN in breath. Gavage administration of MAN to rats causes olfactory epithelial damage and liver enlargement. It was hypothesized that treatment of rats with MAN may result in differential expression of CYP2E1 in tissues leading to tissue-specific toxicity via increased in situ formation of cytotoxic MAN metabolites. In this study, male F344 rats received 60 mg MAN/kg and were sacrificed 6, 12, or 24 h after a single dose, or 24 h after 7 consecutive daily doses. Liver, lung, and nasal tissues were collected. Reverse-transcription polymerase chain reaction (RT-PCR), Western blotting, and immunohistochemical staining were used to assess CYP2E1 expression and localization, and chlorzoxazone hydroxylation was used as a measure of CYP2E1 catalytic activity. Present results showed that CYP2E1 mRNA was increased in lung and nasal tissues with minimal effects in liver of MAN-treated rats. Induction of CYP2E1 protein expression was detected in lung. CYP2E1 activity was higher in liver and lung microsomes from MAN-treated rats when compared to control animals. To compare the effects of MAN and acetone, male F344 rats received a single acetone dose (5 ml/kg) by gavage. After 12 h, acetone treatment resulted in a significant increase in the levels of CYP2E1 mRNA and protein in lung and nasal tissues, with no obvious change noted in the liver. Overall, these data suggest that administration of MAN to rats causes increased expression of CYP2E1 in lung, liver, and nasal tissues. These results also show that acetone induces the expression of CYP2E1 at both the mRNA and protein levels in rat nasal and lung tissues. In conclusion, MAN increased the expression of CYP2E1, and this effect varied as a function of time, length of exposure, and tissue examined. While the damage in the olfactory mucosa due to MAN treatment may not be explained by the observed induction of CYP2E1, it is possible that other CYPs may play a role in the in situ bioactivation of MAN.