Merck
  • Norepinephrine metabolism in neuron: dissociation between 3,4-dihydroxyphenylglycol and 3,4-dihydroxymandelic acid pathways.
Norepinephrine metabolism in neuron: dissociation between 3,4-dihydroxyphenylglycol and 3,4-dihydroxymandelic acid pathways.
Acta pharmacologica Sinica (2002-02-28)
Wen-Xin Dong, Xiang-Lian Ni
ABSTRACT
To investigate the pre-synaptic metabolism of norepinephrine (NE), judged by variations in plasma concentration of 3,4-dihydroxyphenylglycol (DHPG) and 3,4-dihydroxymandelic acid (DOMA). Pithed and electrically stimulated (2.5 Hz) rats were given intravenous infusion of exogenous NE (6 nmol . kg-1 . min-1). Plasma NE, DHPG, DOMA, and the activities of mono- amine oxidases (MAO) were measured with the radio-enzymatic assay. Exogenous NE induces an about 100-fold increase in plasma NE concentration while blood pressure remained within normal limits. A 12-fold increase in plasma DHPG and 1.2-fold increase for DOMA were observed. When NE transportation across the pre-synaptic membrane was inhibited by desipramine (2 mg/kg, iv), a great reduction in plasma DHPG concentration (about 25 % of control) was observed while DOMA remained unchanged. When MAO-A activity was inhibited to 25 % of control by clorgyline (2 mg/kg, iv) and MAO-B to 30 % by deprenyl, the plasma DHPG and DOMA concentrations were reduced to 15 % and 70 % of controls, and to 26 % and 76 % of controls, respectively. When clorgyline and deprenyl were combined, plasma DHPG was vanished (less than 2 % of control) while plasma DOMA remained in the same range (72 % of control). The metabolizing system of NE in pre-synapse, associating with the pre-synaptic reuptake plus oxidative deamination on the external membrane of mitochondria, is predominant for the reduction to DHPG.
Sigma-Aldrich
DL-3,4-Dihydroxymandelic acid, 95%