Merck
  • Spectroscopic study of the interactions of alkali fluorides with D-xylose.
Spectroscopic study of the interactions of alkali fluorides with D-xylose.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2002-01-05)
J Fernandez-Bertrán, E Reguera, P Ortiz
ABSTRACT
The interactions of alkali fluorides with D-xylose have been studied by X-ray diffraction (XRD), infrared spectroscopy (IR), nuclear magnetic resonance (NMR, 1H and 13C) and atomic absorption spectrophotometry. KF and CsF form complexes with D-xylose in a 1:1 molar ratio. These complexes can be obtained by solid state milling the reactants in an agate mortar or from methanolic solutions of the sugar and the salt. LiF and NaF do not form complex with D-xylose. IR and XRD prove the identical nature of the complexes obtained by milling and from solution. IR spectra indicate strong perturbation of the OH stretching vibrations with considerable shifts to lower frequencies, which must be caused by strong hydrogen bond formation to the fluorine anion. The perturbations of C-O bond are weak, indicating that cation binding to the oxygen atoms is not the main interaction responsible for the complex formation. 1H NMR spectra of the D-xylose-KF complex dissolved in deuterium oxide is equal to that of pure D-xylose, indicating the destruction of the complex in solution. The complex is stable in DMSO, and 13C spectra of the complex in DMSO-d6 and in solid state (CPMAS) spectra are in accordance with the observed interactions in the IR spectra. As far as we know, this is the first report of a sugar-halide salt complex in which the anion instead of the cation provides the binding forces.
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis