Merck
  • Phosphorylation and metabolism of sucrose and its five linkage-isomeric alpha-D-glucosyl-D-fructoses by Klebsiella pneumoniae.
Phosphorylation and metabolism of sucrose and its five linkage-isomeric alpha-D-glucosyl-D-fructoses by Klebsiella pneumoniae.
Carbohydrate research (2001-04-27)
J Thompson, S A Robrish, A Pikis, A Brust, F W Lichtenthaler
ABSTRACT
Not only sucrose but the five isomeric alpha-D-glucosyl-D-fructoses trehalulose, turanose, maltulose, leucrose, and palatinose are utilized by Klebsiella pneumoniae as energy sources for growth, thereby undergoing phosphorylation by a phosphoenolpyruvate-dependent phosphotransferase system uniformly at 0-6 of the glucosyl moiety. Similarly, maltose, isomaltose, and maltitol, when exposed to these conditions, are phosphorylated regiospecifically at O-6 of their non-reducing glucose portion. The structures of these novel compounds have been established unequivocally by enzymatic analysis, acid hydrolysis, FAB negative-ion spectrometry, and 1H and 13C NMR spectroscopy. In cells of K. pneumoniae, hydrolysis of sucrose 6-phosphate is catalyzed by sucrose 6-phosphate hydrolase from Family 32 of the glycosylhydrolase superfamily. The five 6'-O-phosphorylated alpha-D-glucosyl-fructoses are hydrolyzed by an inducible (approximately 49-50 Kda) phospho-alpha-glucosidase from Family 4 of the glycosylhydrolase superfamily.
Sigma-Aldrich
D-(+)-Turanose, ≥98%