Merck
  • A labeling, detection, and purification system based on 4-hydroxyazobenzene-2-carboxylic acid: an extension of the avidin-biotin system.
A labeling, detection, and purification system based on 4-hydroxyazobenzene-2-carboxylic acid: an extension of the avidin-biotin system.
Analytical biochemistry (2000-08-31)
H Hofstetter, M Morpurgo, O Hofstetter, E A Bayer, M Wilchek
ABSTRACT
We introduce a new nonradioactive, chromogenic label based on 4-hydroxyazobenzene-2-carboxylic acid (HABA), which is suitable for bioanalytical application, e.g., detection, localization, isolation, and purification. The HABA label is superior to other systems where it is difficult to separate labeled from unlabeled molecules or to determine the amount of label. HABA is readily detected spectroscopically by its absorption at 350 nm or by its interaction with avidin that results in a red shift to 500 nm. The HABA reagents described can be conjugated to a variety of functional groups on biomolecules and purified thereafter by affinity chromatography on an avidin column. The interaction of the HABAylated biomolecules with their corresponding targets is detected with high-affinity anti-HABA antibodies or with avidin. The nonradioactive, chromogenic HABA-based reagents form a homogeneous system that can complement or replace systems where facile quantification of the label is desired.
Sigma-Aldrich
2-(4-Hydroxyphenylazo)benzoic acid, ≥90% (HPLC)