Merck
  • Histomorphometry and quantification of nucleolar organizer regions in bovine thyroid containing methylthiouracil residues.
Histomorphometry and quantification of nucleolar organizer regions in bovine thyroid containing methylthiouracil residues.
Veterinary pathology (1999-11-24)
R Serakides, V A Nunes, R L Santos, G D Cassali, P P Costa Neto
ABSTRACT
In order to study the morphology and morphometry and to characterize and quantify the nucleolar organizer regions (NORs) of bovine thyroids containing methylthiouracil (MTU) residues, five animals were orally treated with a suspension of MTU (5 g/animal/day) for 20 days (group A). This treatment protocol was interrupted 5 days before the the animals were slaughtered. Six animals receiving placebos composed group B. A third group (group C) was composed of normal thyroids obtained from a slaughterhouse. All glands were previously assessed for detection of antithyroid residues by chromatography, and only those glands from MTU-treated animals were positive. Follicles of glands from group A showed wide variation in size and shape. There was a predominance of small follicles covered by multiple layers of columnar cells, sometimes forming papillary projections into the lumen, characterizing severe interfollicular and intrafollicular adenomatosis. Many follicles had vacuolated cells with nuclei showing karyolysis or pyknosis and reduced amounts of a low-density and very excavated colloid. They also showed higher follicular epithelia and larger proportions of their structural components when compared with glands of groups B and C. In the thyroids from group A, the argyrophilic nucleolar organizer region-associated proteins (AgNORs) were greater in number, with small ones scattered all over the nucleus. Although the size of AgNORs in thyroids from groups B and C was variable, these AgNORs were fewer and larger than were those in glands from group A. In conclusion, the MTU induces proliferation and regressive changes in follicular cells, and the AgNOR technique is efficient to distinguish different degrees of thyroid hyperplasia.
Sigma-Aldrich
6-Methyl-2-thiouracil, purum, ≥98.0% S basis (elemental analysis)
Supelco
6-Methyl-2-thiouracil, VETRANAL®, analytical standard