Merck
  • In situ generation of Fmoc-amino acid chlorides using bis-(trichloromethyl) carbonate and its utilization for difficult couplings in solid-phase peptide synthesis.
In situ generation of Fmoc-amino acid chlorides using bis-(trichloromethyl) carbonate and its utilization for difficult couplings in solid-phase peptide synthesis.
The journal of peptide research : official journal of the American Peptide Society (1999-07-29)
E Falb, T Yechezkel, Y Salitra, C Gilon
ABSTRACT
This paper reports procedures for the straightforward in situ generation of Fmoc-amino acid chlorides using bis-(trichloromethyl)carbonate (BTC) and their utilization for difficult couplings during solid-phase peptide synthesis. The BTC-mediated coupling of all Fmoc-protected proteinogenic amino acids to a large variety of N-alkylated amino acid-peptidyl-resin was studied. The majority of the couplings proceeded with quantitative conversion and without racemization. The utilization of BTC-mediated coupling for facile solid-phase synthesis of backbone cyclic peptides is presented.
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triphosgene, reagent grade, 98%