Merck
  • Posttranslational N-myristoylation of BID as a molecular switch for targeting mitochondria and apoptosis.
Posttranslational N-myristoylation of BID as a molecular switch for targeting mitochondria and apoptosis.
Science (New York, N.Y.) (2000-12-02)
J Zha, S Weiler, K J Oh, M C Wei, S J Korsmeyer
ABSTRACT
Many apoptotic molecules relocate subcellularly in cells undergoing apoptosis. The pro-apoptotic protein BID underwent posttranslational (rather than classic cotranslational) N-myristoylation when cleavage by caspase 8 caused exposure of a glycine residue. N-myristoylation enabled the targeting of a complex of p7 and myristoylated p15 fragments of BID to artificial membranes bearing the lipid composition of mitochondria, as well as to intact mitochondria. This post-proteolytic N-myristoylation serves as an activating switch, enhancing BID-induced release of cytochrome c and cell death.
Sigma-Aldrich
BID human, ≥95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
BID, Caspase-8-cleaved from mouse, ≥95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution