Merck
  • Validation of an LC-MS method for the detection and quantification of BZP and TFMPP and their hydroxylated metabolites in human plasma and its application to the pharmacokinetic study of TFMPP in humans.
Validation of an LC-MS method for the detection and quantification of BZP and TFMPP and their hydroxylated metabolites in human plasma and its application to the pharmacokinetic study of TFMPP in humans.
Journal of forensic sciences (2010-06-11)
Ushtana Antia, Malcolm D Tingle, Bruce R Russell
ABSTRACT
An LC-MS method was developed for benzylpiperazine (BZP) and trifluoromethylphenylpiperazine (TFMPP), constituents of "party pills" or "legal herbal highs," and their metabolites in human blood plasma. Compounds were resolved using a mixture of ammonium formate (pH 4.5, 0.01 M) and acetonitrile (flow rate of 1.0 mL/min) with a C18 column. Calibration curves were linear from 1 to 50 ng/mL (R(2) > 0.99); the lower limit of quantification (LLOQ) was 5 ng/mL; the accuracy was >90%; the intra- and interday relative standard deviations (R.S.D) were <5% and <10%, respectively. Human plasma concentrations of TFMPP were measured in blood samples taken from healthy adults (n = 6) over 24 h following a 60-mg oral dose of TFMPP: these peaked at 24.10 ng/mL (±1.8 ng/mL) (C(max) ) after 90 min (T(max)). Plasma concentrations of 1-(3-trifluoromethyl-4-hydroxyphenyl) piperazine peaked at 20.2 ng/mL (±4.6 ng/mL) after 90 min. TFMPP had two disposition phases (t(½) = 2.04 h (±0.19 h) and 5.95 h (±1.63 h). Apparent clearance (Cl/F) was 384 L/h (±45 L/h).
Product Description
Supelco
Benzyl piperazine dihydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®