- Lipemia retinalis: case report and review of the literature.
Lipemia retinalis: case report and review of the literature.
Journal of AAPOS : the official publication of the American Association for Pediatric Ophthalmology and Strabismus (2013-01-23)
Alon Zahavi, Moshe Snir, Yonina Ron Kella
PMID23337350
ABSTRACT
We report the case of a 12-week-old boy presenting with increased cholesterol and triglyceride levels. Examination revealed lipemia retinalis. Genetic evaluation demonstrated lipoprotein lipase deficiency. The patient was treated with dietary restrictions, which resulted in rapid clinical improvement.
Sigma-Aldrich
Lipoprotein Lipase from bovine milk, ammonium sulfate suspension, ≥2,000 units/mg protein (BCA)
Sigma-Aldrich
Lipoprotein Lipase from Pseudomonas sp., lyophilized, powder, yellow-brown, ≥160 U/mg
Sigma-Aldrich
Lipoprotein Lipase from Burkholderia sp., lyophilized powder, ≥50,000 units/mg solid