Merck
  • Genome-wide mapping of nucleosome occupancy, histone modifications, and gene expression using next-generation sequencing technology.
Genome-wide mapping of nucleosome occupancy, histone modifications, and gene expression using next-generation sequencing technology.
Methods in enzymology (2012-08-30)
Gang Wei, Gangqing Hu, Kairong Cui, Keji Zhao
ABSTRACT
Gene transcription can be regulated through alteration of chromatin structure, such as changes in nucleosome positioning and histone-modification patterns. Recent development of techniques based on the next-generation sequencing technology has allowed high-resolution analysis of genome-wide distribution of these chromatin features. In this chapter, we describe in detail the protocols of ChIP-Seq and MNase-Seq, which have been developed to detect the genome-wide profiles of transcription factor binding, histone modifications, and nucleosome occupancy. We also describe RNA-Seq protocols used to map global gene expression profiles.
Sigma-Aldrich
Nuclease micrococcal from Staphylococcus aureus, 100-300 units/mg protein
Sigma-Aldrich
Nuclease micrococcal from Staphylococcus aureus, 100-300 units/mg protein