Merck
  • Monoclonal anti-HMGB1 (high mobility group box chromosomal protein 1) antibody protection in two experimental arthritis models.
Monoclonal anti-HMGB1 (high mobility group box chromosomal protein 1) antibody protection in two experimental arthritis models.
Molecular medicine (Cambridge, Mass.) (2011-06-15)
Hanna Schierbeck, Peter Lundbäck, Karin Palmblad, Lena Klevenvall, Helena Erlandsson-Harris, Ulf Andersson, Lars Ottosson
ABSTRACT
High mobility group box chromosomal protein 1 (HMGB1) is a DNA-binding nuclear protein that can be released from dying cells and activated myeloid cells. Extracellularly, HMGB1 promotes inflammation. Experimental studies demonstrate HMGB1 to be a pathogenic factor in many inflammatory conditions including arthritis. HMGB1-blocking therapies in arthritis models alleviate disease and confer significant protection against cartilage and bone destruction. So far, the most successful HMGB1-targeted therapies have been demonstrated with HMGB1-specific polyclonal antibodies and with recombinant A box protein, a fragment of HMGB1. The present study is the first to evaluate the potential of a monoclonal anti-HMGB1 antibody (2G7, mouse IgG2b) to ameliorate arthritis. Effects of repeated injections of this antibody have now been studied in two conceptually different models of arthritis: collagen type II-induced arthritis (CIA) in DBA/1 mice and in a spontaneous arthritis disease in mice with combined deficiencies for genes encoding for the enzyme DNase type II and interferon type I receptors. These mice are unable to degrade phagocytozed DNA in macrophages and develop chronic, destructive polyarthritis. Therapeutic intervention in CIA and prophylactic administration of anti-HMGB1 monoclonal antibody (mAb) in the spontaneous arthritis model significantly ameliorated the clinical courses. Anti-HMGB1 mAb therapy also partially prevented joint destruction, as demonstrated by histological examination. The beneficial antiarthritic effects by the anti-HMGB1 mAb in two diverse models of arthritis represent additional proof-of-concept, indicating that HMGB1 may be a valid target molecule to consider for development of future clinical therapy.
Sigma-Aldrich
Deoxyribonuclease II from porcine spleen, Type IV, lyophilized powder, 2,000-6,000 Kunitz units/mg protein (biuret)
Sigma-Aldrich
Deoxyribonuclease II from bovine spleen, Type V, essentially salt-free, lyophilized powder, ≥1,000 units/mg protein