Merck
  • An amperometric biosensor based on ascorbate oxidase immobilized in poly(3,4-ethylenedioxythiophene)/multi-walled carbon nanotubes composite films for the determination of L-ascorbic acid.
An amperometric biosensor based on ascorbate oxidase immobilized in poly(3,4-ethylenedioxythiophene)/multi-walled carbon nanotubes composite films for the determination of L-ascorbic acid.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2011-05-12)
Ming Liu, Yangping Wen, Jingkun Xu, Haohua He, Dong Li, Ruirui Yue, Guodong Liu
ABSTRACT
An amperometric L-ascorbic acid (AA) biosensor fabricated by immobilizing ascorbate oxidase (AO) in poly(3,4-ethylenedioxythiophene) (PEDOT) and multi-walled carbon nanotubes (MWCNTs) composite films was reported for the first time. The entrapment of AO in PEDOT/MWCNTs composite films was performed during an electrochemical polymerization process. The influence of various experimental conditions was examined for determining the optimum analytical performance. The response of the biosensor towards AA under the optimized conditions is linear from 0.05 to 20 mM with a detection limit of 15 µM (S/N = 3). The biosensor shows a response time of 20 s and a sensitivity of 23.95 mA M(-1) cm(-2). The apparent Michaelis-Menten constant (K(m)) and apparent activation energy (E(a)) are 19.5 mM and 21 kJ mol(-1), respectively. Moreover, the biosensor exhibits good anti-interferent ability, good reproducibility and remarkable storage stability.
Sigma-Aldrich
Ascorbate Oxidase from Cucurbita sp., lyophilized powder, 1,000-3,000 units/mg protein
Pricing and availability is not currently available.