Merck
Endo-beta-galactosidases and keratanase.
Current protocols in molecular biology (2008-02-12)
M N Fukuda
ABSTRACT
This overview covers the endo-beta-galactosidases; enzyme is capable of hydrolyzing a wide range of glycoconjugates. Endo-beta-galactosidases from numerous sources are discussed in terms of their substrate specificities and substrates, as well as their practical research applications.