Merck
  • Effective amplification of long targets from cloned inserts and human genomic DNA.
Effective amplification of long targets from cloned inserts and human genomic DNA.
Proceedings of the National Academy of Sciences of the United States of America (1994-06-07)
S Cheng, C Fockler, W M Barnes, R Higuchi
ABSTRACT
We have used the polymerase chain reaction (PCR) to amplify up to 22 kb of the beta-globin gene cluster from human genomic DNA and up to 42 kb from phaga lambda DNA. We have also amplified 91 human genomic inserts of 9-23 kb directly from recombinant lambda plaques. To do this, we increased pH, added glycerol and dimethyl sulfoxide, decreased denaturation times, increased extension times, and used a secondary thermostable DNA polymerase that possesses a 3'-to 5'-exonuclease, or "proofreading," activity. Our "long PCR" protocols maintain the specificity required for targets in genomic DNA by using lower levels of polymerase and temperature and salt conditions for specific primer annealing. The ability to amplify DNA sequences of 10-40 kb will bring the speed and simplicity of PCR to genomic mapping and sequencing and facilitate studies in molecular genetics.
Sigma-Aldrich
JumpStart AccuTaq LA DNA Polymerase, Hot-start high fidelity Taq enzyme, 10X buffer included
Sigma-Aldrich
REDAccuTaq® LA DNA Polymerase, High fidelity Taq with inert dye, 10X buffer included
Sigma-Aldrich
AccuTaq LA DNA Polymerase, High fidelity Taq enzyme, with 10X buffer & DMSO
Sigma-Aldrich
JumpStart REDAccuTaq® LA DNA Polymerase, Long and accurate hot-start Taq with inert dye, 10X buffer included
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent