Merck
  • Disruption of ZNF334 promotes triple-negative breast carcinoma malignancy through the SFRP1/ Wnt/β-catenin signaling axis.
Disruption of ZNF334 promotes triple-negative breast carcinoma malignancy through the SFRP1/ Wnt/β-catenin signaling axis.
Cellular and molecular life sciences : CMLS (2022-05-05)
Zhaobo Cheng, Renjie Yu, Li Li, Junhao Mu, Yijia Gong, Fan Wu, Yujia Liu, Xiangyi Zhou, Xiaohua Zeng, Yongzhong Wu, Ran Sun, Tingxiu Xiang
ABSTRACT
Zinc-finger proteins (ZNFs) constitute the largest transcription factor family in the human genome. The family functions in many important biological processes involved in tumorigenesis. In our research, we identified ZNF334 as a novel tumor suppressor of triple-negative breast cancer (TNBC). ZNF334 expression was usually reduced in breast cancerv (BrCa) tissues and TNBC cell lines MDA-MB-231 (MB231) and YCCB1. We observed that promoter hypermethylation of ZNF334 was common in BrCa cell lines and tissues, which was likely responsible for its reduced expression. Ectopic expression of ZNF334 in TNBC cell lines MB231 and YCCB1 could suppress their growth and metastatic capacity both in vitro and in vivo, and as well induce cell cycle arrest at S phase and cell apoptosis. Moreover, re-expression of ZNF334 in TNBC cell lines could rescue Epithelial-Mesenchymal Transition (EMT) process and restrain stemness, due to up-regulation of SFRP1, which is an antagonist of Wnt/β-catenin signaling. In conclusion, we verified that ZNF334 had a suppressive function of TNBC cell lines by targeting the SFRP1/Wnt/β-catenin signaling axis, which might have the potentials to become a new biomarker for diagnosis and treatment of TNBC patients.
Sigma-Aldrich
Anti-ZNF334 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)