Merck
  • Regulation of the intracellular localization of Foxo3a by stress-activated protein kinase signaling pathways in skeletal muscle cells.
Regulation of the intracellular localization of Foxo3a by stress-activated protein kinase signaling pathways in skeletal muscle cells.
Molecular and cellular biology (2009-11-18)
Stephan Clavel, Sandrine Siffroi-Fernandez, Anne Sophie Coldefy, Kim Boulukos, Didier F Pisani, Benoît Dérijard
ABSTRACT
Muscle atrophy is a debilitating process associated with many chronic wasting diseases, like cancer, diabetes, sepsis, and renal failure. Rapid loss of muscle mass occurs mainly through the activation of protein breakdown by the ubiquitin proteasome pathway. Foxo3a transcription factor is critical for muscle atrophy, since it activates the expression of ubiquitin ligase Atrogin-1. In several models of atrophy, inhibition of the phosphatidylinositol 3-kinase (PI3K)/Akt signaling pathway induces nuclear import of Foxo3a through an Akt-dependent process. This study aimed to identify signaling pathways involved in the control of Foxo3a nuclear translocation in muscle cells. We observed that after nuclear import of Foxo3a by PI3K/Akt pathway inhibition, activation of stress-activated protein kinase (SAPK) pathways induced nuclear export of Foxo3a through CRM1. This mechanism involved the c-Jun NH(2)-terminal kinase (JNK) signaling pathway and was independent of Akt. Likewise, we showed that inhibition of p38 induced a massive nuclear relocalization of Foxo3a. Our results thus suggest that SAPKs are involved in the control of Foxo3a nucleocytoplasmic translocation in C2C12 cells. Moreover, activation of SAPKs decreases the expression of Atrogin-1, and stable C2C12 myotubes, in which the p38 pathway is constitutively activated, present partial protection against atrophy.
Product Description
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
SKU
Pack Size
Availability
Price
Quantity
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O