Merck
  • Identification of druggable host dependency factors shared by multiple SARS-CoV-2 variants of concern.
Identification of druggable host dependency factors shared by multiple SARS-CoV-2 variants of concern.
Journal of molecular cell biology (2024-02-02)
Ilaria Frasson, Linda Diamante, Manuela Zangrossi, Elena Carbognin, Anna Dalla Pietà, Alessandro Penna, Antonio Rosato, Ranieri Verin, Filippo Torrigiani, Cristiano Salata, Marìa Paula Dizanzo, Lorenzo Vaccaro, Davide Cacchiarelli, Sara N Richter, Marco Montagner, Graziano Martello
ABSTRACT
The high mutation rate of SARS-CoV-2 leads to the emergence of multiple variants, some of which are resistant to vaccines and drugs targeting viral elements. Targeting host dependency factors, e.g. cellular proteins required for viral replication, would help prevent the development of resistance. However, it remains unclear whether different SARS-CoV-2 variants induce conserved cellular responses and exploit the same core host factors. To this end, we compared three variants of concern and found that the host transcriptional response was conserved, differing only in kinetics and magnitude. Clustered regularly interspaced short palindromic repeats screening identified host genes required for each variant during infection. Most of the genes were shared by multiple variants. We validated our hits with small molecules and repurposed the US Food and Drug Administration-approved drugs. All the drugs were highly active against all the tested variants, including new variants that emerged during the study (Delta and Omicron). Mechanistically, we identified reactive oxygen species production as a key step in early viral replication. Antioxidants such as N-acetyl cysteine (NAC) were effective against all the variants in both human lung cells and a humanized mouse model. Our study supports the use of available antioxidant drugs, such as NAC, as a general and effective anti-COVID-19 approach.
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Syk Inhibitor IV, BAY 61-3606, The Syk Inhibitor IV, BAY 61-3606, also referenced under CAS 732938-37-8, controls the biological activity of Syk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.