MilliporeSigma
  • Automated immunohistochemical method to analyze large areas of the human cortex.
Automated immunohistochemical method to analyze large areas of the human cortex.
Journal of neuroscience methods (2017-11-12)
Mohamad Abbass, Kathleen Trought, David Long, Anton Semechko, Albert H C Wong
ABSTRACT
There have been inconsistencies in the histological abnormalities found in the cerebral cortex from patients with schizophrenia, bipolar disorder and major depression. Discrepancies in previously published reports may arise from small sample sizes, inconsistent methodology and biased cell counting. We applied automated quantification of neuron density, neuron size and cortical layer thickness in large regions of the cerebral cortex in psychiatric patients. This method accurately segments DAPI positive cells that are also stained with CUX2 and FEZF2. Cortical layer thickness, neuron density and neuron size were automatically computed for each cortical layer in numerous Brodmann areas. We did not find pronounced cytoarchitectural abnormalities in the anterior cingulate cortex or orbitofrontal cortex in patients with schizophrenia, bipolar disorder or major depressive disorder. There were no significant differences in layer thickness measured in immunohistochemically stained slides compared with traditional Nissl stained slides. Automated cell counts were correlated, reliable and consistent with manual counts, while being much less time-consuming. We demonstrate the validity of using a novel automated analysis approach to post-mortem brain tissue. We were able to analyze large cortical areas and quantify specific cell populations using immunohistochemical markers. Future analyses could benefit from efficient automated analysis.
Sigma-Aldrich
Monoclonal Anti-CUX2 antibody produced in mouse, clone 2H8, purified immunoglobulin, buffered aqueous solution