Merck
  • The midnolin-proteasome pathway catches proteins for ubiquitination-independent degradation.
The midnolin-proteasome pathway catches proteins for ubiquitination-independent degradation.
Science (New York, N.Y.) (2023-08-24)
Xin Gu, Christopher Nardone, Nolan Kamitaki, Aoyue Mao, Stephen J Elledge, Michael E Greenberg
ABSTRACT
Cells use ubiquitin to mark proteins for proteasomal degradation. Although the proteasome also eliminates proteins that are not ubiquitinated, how this occurs mechanistically is unclear. Here, we found that midnolin promoted the destruction of many nuclear proteins, including transcription factors encoded by the immediate-early genes. Diverse stimuli induced midnolin, and its overexpression was sufficient to cause the degradation of its targets by a mechanism that did not require ubiquitination. Instead, midnolin associated with the proteasome via an α helix, used its Catch domain to bind a region within substrates that can form a β strand, and used a ubiquitin-like domain to promote substrate destruction. Thus, midnolin contains three regions that function in concert to target a large set of nuclear proteins to the proteasome for degradation.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-C11orf84 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution