Merck
  • Inhibition of DPAGT1 suppresses HER2 shedding and trastuzumab resistance in human breast cancer.
Inhibition of DPAGT1 suppresses HER2 shedding and trastuzumab resistance in human breast cancer.
The Journal of clinical investigation (2023-07-18)
Muwen Yang, Yue Li, Lingzhi Kong, Shumei Huang, Lixin He, Pian Liu, Shuang Mo, Xiuqing Lu, Xi Lin, Yunyun Xiao, Dongni Shi, Xinjian Huang, Boyu Chen, Xiangfu Chen, Ying Ouyang, Jun Li, Chuyong Lin, Libing Song
ABSTRACT
Human epidermal growth factor receptor 2-targeted (HER2-targeted) therapy is the mainstay of treatment for HER2+ breast cancer. However, the proteolytic cleavage of HER2, or HER2 shedding, induces the release of the target epitope at the ectodomain (ECD) and the generation of a constitutively active intracellular fragment (p95HER2), impeding the effectiveness of anti-HER2 therapy. Therefore, identifying key regulators in HER2 shedding might provide promising targetable vulnerabilities against resistance. In the current study, we found that upregulation of dolichyl-phosphate N-acetylglucosaminyltransferase (DPAGT1) sustained high-level HER2 shedding to confer trastuzumab resistance, which was associated with poor clinical outcomes. Upon trastuzumab treatment, the membrane-bound DPAGT1 protein was endocytosed via the caveolae pathway and retrogradely transported to the ER, where DPAGT1 induced N-glycosylation of the sheddase - ADAM metallopeptidase domain 10 (ADAM10) - to ensure its expression, maturation, and activation. N-glycosylation of ADAM10 at N267 protected itself from ER-associated protein degradation and was essential for DPAGT1-mediated HER2 shedding and trastuzumab resistance. Importantly, inhibition of DPAGT1 with tunicamycin acted synergistically with trastuzumab treatment to block HER2 signaling and reverse resistance. These findings reveal a prominent mechanism for HER2 shedding and suggest that targeting DPAGT1 might be a promising strategy against trastuzumab-resistant breast cancer.
Sigma-Aldrich
Anti-DPAGT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ADAM10 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution