Merck
  • Hepatoprotective effect of gentiopicroside in combination with leflunomide and/or methotrexate in arthritic rats.
Hepatoprotective effect of gentiopicroside in combination with leflunomide and/or methotrexate in arthritic rats.
Life sciences (2020-11-02)
Zhijie Wan, He Li, Xiaohan Wu, Haiyun Zhao, Ran Wang, Mengmeng Li, Jing Liu, Qingfeng Liu, Rui Wang, Xiaotian Li
ABSTRACT
This study aimed to examine whether gentiopicroside (GPS) could exert hepatoprotective effects on leflunomide (LEF)- and/or methotrexate (MTX)-treated arthritic rats through anti-inflammatory and antioxidant pathways. We observed the external symptoms of joints, analysed serum indicators, measured haematological parameters and mRNA levels, and performed HE staining. LEF and/or MTX combined with GPS ameliorated oxidative stress by increasing the mRNA levels of the antioxidant gene Nrf2, GCLC, HO-1, and NQO1, increasing the antioxidant enzymes superoxide dismutase (SOD), glutathione (GSH) and catalase (CAT), reducing the oxidant substance malondialdehyde (MDA), reducing the inflammatory response by decreasing the mRNA levels of NF-κB, tumour necrosis factor-α (TNF-α), interleukin-1β (IL-1β), and interleukin-6 (IL-6), and inhibiting the secretion of the pro-inflammatory cytokines TNFα, IL-6, IL-1β and reducing C-reactive protein (CRP), as well as alleviating the external symptoms of arthritis. These results show that GPS plays an antioxidant and anti-inflammatory role in LEF- and/or MTX-treated arthritic rats by affecting the Nrf2 and NF-κB signalling pathways, thus exerting hepatoprotective effects.
Sigma-Aldrich
Gentiopicroside, ≥98% (HPLC)