Merck
  • Triplet Harvesting by a Fluorescent Emitter Using a Phosphorescent Sensitizer for Blue Organic-Light-Emitting Diodes.
Triplet Harvesting by a Fluorescent Emitter Using a Phosphorescent Sensitizer for Blue Organic-Light-Emitting Diodes.
ACS applied materials & interfaces (2018-12-14)
Hyun-Gu Kim, Hyun Shin, Yeon Hee Ha, Ran Kim, Soon-Ki Kwon, Yun-Hi Kim, Jang-Joo Kim
ABSTRACT
The development of highly efficient blue organic light-emitting diodes (OLEDs) with good stability is currently the most important issue in OLED displays and lighting. This paper reports an efficient blue fluorescent OLED based on a deep-blue-emitting phosphorescent sensitizer [(dfpysipy)2Ir(mpic)] and a conventional fluorescent emitter (TBPe). Efficient triplet harvesting by the fluorescent emitter occurs in the OLED because of sensitization even though the difference in the emission energy between the phosphorescent and fluorescent emissions was only 0.05 eV. These results clearly demonstrate the potential for realizing highly efficient blue fluorescent OLEDs using phosphorescent sensitizers without requiring ultraviolet-emitting phosphorescent dye.
Sigma-Aldrich
4,4′,4′′-Tris(N-(naphthalen-2-yl)-N-phenyl-amino)-triphenylamine