Merck
  • Expression and purification of soluble adenylyl cyclase from Escherichia coli.
Expression and purification of soluble adenylyl cyclase from Escherichia coli.
Methods in molecular biology (Clifton, N.J.) (2003-09-23)
Jeff A Beeler, Wei-Jen Tang
ABSTRACT
This chapter outlines methods to purify soluble adenylyl cyclase (AC7) expressed in an Escherichia coli (E. coli) heterologous expression system. Guidelines are provided for constructing the expression plasmids, optimizing expression, culturing, and purifying the proteins. Purification requires two chromatographic steps. A histidine tag (H6) is incorporated into the expression vector and utilized for affinity purification on a Ni-NTA column. Subsequently, an anion exchange column is employed to further purify the protein.
Sigma-Aldrich
Ro-20-1724, A cell-permeable, selective inhibitor of cAMP-specific phosphodiesterase (PDE IV; IC₅₀ = 2 µM).
Sigma-Aldrich
Anti-Mouse IgG (γ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥90 %, ≥39,000 units/mg protein