Merck
  • Neutral ceramidase-dependent regulation of macrophage metabolism directs intestinal immune homeostasis and controls enteric infection.
Neutral ceramidase-dependent regulation of macrophage metabolism directs intestinal immune homeostasis and controls enteric infection.
Cell reports (2022-03-31)
Rui Sun, Xuemei Gu, Chao Lei, Liang Chen, Shenghui Chu, Guangzhong Xu, Mark A Doll, Yi Tan, Wenke Feng, Leah Siskind, Craig J McClain, Zhongbin Deng
ABSTRACT
It is not clear how the complex interactions between diet and intestinal immune cells protect the gut from infection. Neutral ceramidase (NcDase) plays a critical role in digesting dietary sphingolipids. We find that NcDase is an essential factor that controls intestinal immune cell dynamics. Mice lacking NcDase have reduced cluster of differentiation (CD) 8αβ+ T cells and interferon (IFN)-γ+ T cells and increased macrophages in the intestine and fail to clear bacteria after Citrobacter rodentium infection. Mechanistically, cellular NcDase or extracellular vesicle (EV)-related NcDase generates sphingosine, which promotes macrophage-driven Th1 immunity. Loss of NcDase influences sphingosine-controlled glycolytic metabolism in macrophages, which regulates the bactericidal activity of macrophages. Importantly, administration of dietary sphingomyelin and genetic deletion or pharmacological inhibition of SphK1 can protect against C. rodentium infection. Our findings demonstrate that sphingosine profoundly alters macrophage glycolytic metabolism, leading to intestinal macrophage activation and T cell polarization, which prevent pathogen colonization of the gut.
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, clone 1D6, clone 1D6, Upstate®, from mouse
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution