Merck
  • Cardiac proteomics reveals sex chromosome-dependent differences between males and females that arise prior to gonad formation.
Cardiac proteomics reveals sex chromosome-dependent differences between males and females that arise prior to gonad formation.
Developmental cell (2021-10-17)
Wei Shi, Xinlei Sheng, Kerry M Dorr, Josiah E Hutton, James I Emerson, Haley A Davies, Tia D Andrade, Lauren K Wasson, Todd M Greco, Yutaka Hashimoto, Joel D Federspiel, Zachary L Robbe, Xuqi Chen, Arthur P Arnold, Ileana M Cristea, Frank L Conlon
ABSTRACT
Sex disparities in cardiac homeostasis and heart disease are well documented, with differences attributed to actions of sex hormones. However, studies have indicated sex chromosomes act outside of the gonads to function without mediation by gonadal hormones. Here, we performed transcriptional and proteomics profiling to define differences between male and female mouse hearts. We demonstrate, contrary to current dogma, cardiac sex disparities are controlled not only by sex hormones but also through a sex-chromosome mechanism. Using Turner syndrome (XO) and Klinefelter (XXY) models, we find the sex-chromosome pathway is established by X-linked gene dosage. We demonstrate cardiac sex disparities occur at the earliest stages of heart formation, a period before gonad formation. Using these datasets, we identify and define a role for alpha-1B-glycoprotein (A1BG), showing loss of A1BG leads to cardiac defects in females, but not males. These studies provide resources for studying sex-biased cardiac disease states.
Sigma-Aldrich
Anti-PBDC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-A1BG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution