Merck
  • Quantitative assays to measure the transport activity of the mitochondrial calcium uniporter in cell lines or Xenopus oocytes.
Quantitative assays to measure the transport activity of the mitochondrial calcium uniporter in cell lines or Xenopus oocytes.
STAR protocols (2021-12-09)
Madison X Rodriguez, Anna M Van Keuren, Ming-Feng Tsai
ABSTRACT
The mitochondrial calcium uniporter, which mediates mitochondrial Ca2+ uptake, regulates key cellular functions, including intracellular Ca2+ signaling, cell-fate determination, and mitochondrial bioenergetics. Here, we describe two complementary strategies to quantify the uniporter's transport activity. First, we detail a mitochondrial Ca2+ radionuclide uptake assay in cultured cell lines. Second, we describe electrophysiological recordings of the uniporter expressed in Xenopus oocytes. These approaches enable a detailed kinetic analysis of the uniporter to link its molecular properties to physiological functions. For complete details on the use and execution of this protocol, please refer to Tsai and Tsai (2018) and Phillips et al. (2019).
Sigma-Aldrich
Ru360, Ru360, is a cell-permeable oxygen-bridged dinuclear ruthenium amine complex. Binds to mitochondria with high affinity (Kd = 340 pM) and blocks Ca2+ uptake into mitochondria in vitro (IC₅₀ = 184 pM).