Merck
  • Serjanic Acid Glycosides from Chenopodium hybridum L. with Good Cytotoxicity and Selectivity Profile against Several Panels of Human Cancer Cell Lines.
Serjanic Acid Glycosides from Chenopodium hybridum L. with Good Cytotoxicity and Selectivity Profile against Several Panels of Human Cancer Cell Lines.
Molecules (Basel, Switzerland) (2021-08-28)
Karolina Grabowska, Łukasz Pecio, Agnieszka Galanty, Paweł Żmudzki, Wiesław Oleszek, Irma Podolak
ABSTRACT
Two triterpene saponins, including a novel serjanic acid derivative, were isolated from Chenopodium hybridum L. (Amaranthaceae) aerial parts. Their structures were elucidated by a combination of spectroscopic methods (MS, 1D and 2D NMR). Both compounds were evaluated for cytotoxicity and selectivity on skin, prostate, gastrointestinal, thyroid and lung cancer cells. Their effect was dose and time-dependent with varied potency, the highest against prostate PC3 and melanoma WM793, where IC50 was lower than the reference drug doxorubicin. Structure-activity relationship is briefly discussed.
Product Description
FTC-133 Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.