Merck
  • Niacin treatment of stroke increases synaptic plasticity and axon growth in rats.
Niacin treatment of stroke increases synaptic plasticity and axon growth in rats.
Stroke (2010-07-31)
Xu Cui, Michael Chopp, Alex Zacharek, Cynthia Roberts, Benjamin Buller, Madalina Ion, Jieli Chen
ABSTRACT
Niacin is the most effective medication in current clinical use for increasing high-density lipoprotein cholesterol. We tested the hypothesis that niacin treatment of stroke promotes synaptic plasticity and axon growth in the ischemic brain. Male Wistar rats were subjected to 2 hours of middle cerebral artery occlusion and treated with or without Niaspan (a prolonged-release formulation of niacin, 40 mg/kg) daily for 14 days starting 24 hours after middle cerebral artery occlusion. The expression of synaptophysin, Nogo receptor, Bielschowsky silver, brain-derived neurotrophic factor, and its receptor tropomyosin-related kinase B were measured by immunohistostaining and Western blot, respectively, in the ischemic brain. Complementing in vivo studies, primary cultured neurons were used to test the effect of niacin and high-density lipoprotein on neurite outgrowth and brain-derived neurotrophic factor/tropomyosin-related kinase B expression. Niaspan treatment of stroke significantly increased synaptophysin, Bielschowsky silver, brain-derived neurotrophic factor/tropomyosin-related kinase B expression, and decreased Nogo receptor expression in the ischemic brain compared with middle cerebral artery occlusion control animals (P<0.05, n=8/group). Niacin and high-density lipoprotein treatment significantly increased neurite outgrowth and brain-derived neurotrophic factor/tropomyosin-related kinase B expression in primary cultured neurons. Tropomyosin-related kinase B inhibitor attenuated niacin-induced neurite outgrowth (P<0.05, n=6/group). Niacin treatment of stroke promotes synaptic plasticity and axon growth, which is mediated, at least partially, by the brain-derived neurotrophic factor/tropomyosin-related kinase B pathways.
Sigma-Aldrich
Lipoproteins, High Density, Human Plasma