Merck
  • The phytochemical p-hydroxycinnamic acid suppresses the growth and stimulates the death in human liver cancer HepG2 cells.
The phytochemical p-hydroxycinnamic acid suppresses the growth and stimulates the death in human liver cancer HepG2 cells.
Anti-cancer drugs (2021-02-18)
Masayoshi Yamaguchi, Tomiyasu Murata, Joe W Ramos
ABSTRACT
Hepatocellular carcinoma (HCC) is one of the most prevalent malignant diseases and causes a third of cancer-related death. The prognosis and effective treatment of advanced HCC remains poor in spite of the development of novel therapeutic strategies. In the present study, we investigate anticancer effects of the botanical molecule p-hydroxycinnamic  acid (HCA) in the HepG2 liver cancer model in vitro. Culturing with HCA (10-1000 nM) suppressed colony formation and growth of HepG2 cells. Mechanistically, culturing with HCA decreased levels of Ras, PI3K, Akt, MAPK, NF-κB p65 and β-catenin, which are linked to processes of cell signaling and transcription, and increased levels of retinoblastoma and regucalcin, which are suppressors for carcinogenesis. These alterations may lead to the suppression of cell growth. Furthermore, culturing with HCA (10-1000 nM) stimulated cell death due to increased caspase-3 levels. Interestingly, the effects of HCA on the growth and death of HepG2 cells were inhibited by culturing with CH223191, an antagonist of aryl hydrocarbon receptor (AHR), suggesting that the flavonoid effects are, at least partly, mediated by activation of AHR signaling. Notably, HCA blocked stimulatory effects of Bay K 8644, an agonist of L-type calcium channel, on the growth of HepG2 cells. Thus, our study demonstrates that HCA suppresses the growth and stimulates the death of human liver cancer HepG2 cells in vitro. The botanical molecule HCA may therefore be a useful tool in the treatment of HCC, providing a novel strategy for the therapy of human liver cancers.
Sigma-Aldrich
Anti-RGN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1