Merck
  • Caspase-4/11 exacerbates disease severity in SARS-CoV-2 infection by promoting inflammation and immunothrombosis.
Caspase-4/11 exacerbates disease severity in SARS-CoV-2 infection by promoting inflammation and immunothrombosis.
Proceedings of the National Academy of Sciences of the United States of America (2022-05-20)
Mostafa M Eltobgy, Ashley Zani, Adam D Kenney, Shady Estfanous, Eunsoo Kim, Asmaa Badr, Cierra Carafice, Kylene Daily, Owen Whitham, Maciej Pietrzak, Amy Webb, Jeffrey Kawahara, Adrian C Eddy, Parker Denz, Mijia Lu, Mahesh Kc, Mark E Peeples, Jianrong Li, Jian Zhu, Jianwen Que, Richard Robinson, Oscar Rosas Mejia, Rachael E Rayner, Luanne Hall-Stoodley, Stephanie Seveau, Mikhail A Gavrilin, Xiaoli Zhang, Jeronay Thomas, Jacob E Kohlmeier, Mehul S Suthar, Eugene Oltz, Andrea Tedeschi, Frank H Robledo-Avila, Santiago Partida-Sanchez, Emily A Hemann, Eman Abdelrazik, Adriana Forero, Shahid M Nimjee, Prosper N Boyaka, Estelle Cormet-Boyaka, Jacob S Yount, Amal O Amer
ABSTRACT
Severe acute respiratory syndrome coronavirus 2 (SARS–CoV-2) is a worldwide health concern, and new treatment strategies are needed. Targeting inflammatory innate immunity pathways holds therapeutic promise, but effective molecular targets remain elusive. Here, we show that human caspase-4 (CASP4) and its mouse homolog, caspase-11 (CASP11), are up-regulated in SARS–CoV-2 infections and that CASP4 expression correlates with severity of SARS–CoV-2 infection in humans. SARS–CoV-2–infected Casp11−/− mice were protected from severe weight loss and lung pathology, including blood vessel damage, compared to wild-type (WT) mice and mice lacking the caspase downstream effector gasdermin-D (Gsdmd−/−). Notably, viral titers were similar regardless of CASP11 knockout. Global transcriptomics of SARS–CoV-2–infected WT, Casp11−/−, and Gsdmd−/− lungs identified restrained expression of inflammatory molecules and altered neutrophil gene signatures in Casp11−/− mice. We confirmed that protein levels of inflammatory mediators interleukin (IL)-1β, IL-6, and CXCL1, as well as neutrophil functions, were reduced in Casp11−/− lungs. Additionally, Casp11−/− lungs accumulated less von Willebrand factor, a marker for endothelial damage, but expressed more Kruppel-Like Factor 2, a transcription factor that maintains vascular integrity. Overall, our results demonstrate that CASP4/11 promotes detrimental SARS–CoV-2–induced inflammation and coagulopathy, largely independently of GSDMD, identifying CASP4/11 as a promising drug target for treatment and prevention of severe COVID-19.
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Albumin, Tetramethylrhodamine isothiocyanate bovine, lyophilized powder