Merck
  • Surface interactions and degradation of a fluoroquinolone antibiotic in the dark in aqueous TiO2 suspensions.
Surface interactions and degradation of a fluoroquinolone antibiotic in the dark in aqueous TiO2 suspensions.
The Science of the total environment (2015-06-19)
Jonathan W Peterson, Baohua Gu, Michael D Seymour
ABSTRACT
Fluoroquinolone antibiotics (FQs) are important drugs used in human and veterinary medicine. Their detection in natural waters and waste water treatment plants, along with increased resistance to FQs among some bacteria, have generated an increased interest in the fate of these drugs in the environment. Partitioning of FQs between an aqueous solution and attendant substrates depends, in part, on the surface reactivity of the adsorbent, commonly a function of particle size, surface charge, and functional groups. This study investigated the surface interactions between the FQ drug ofloxacin (OFL) and titanium oxide (TiO2), a common catalyst and widely-observed constituent in many consumer products. Raman and fluorescence spectroscopic techniques, as well as LC/MS, were used to determine the OFL moieties present on TiO2 surfaces and in attendant solutions. Raman spectra indicate that the CO (ketone) group of the quinolone core, the NH(+) of the piperazinyl ring, and CH3 of benzoxazine core are the most active in sorption onto the TiO2 surface. Raman spectra also show that the sorbed benzoxazine-quinolone core and piperazinyl moieties are readily desorbed from the surface by re-suspending samples in water. Importantly, we found that OFL could be degraded by reacting with TiO2 even in the dark. Complementary LC/MS analysis of the attendant supernatants indicates the presence of de-piperazinylated and de-carboxylated OFL breakdown products in supernatant solutions. Together, both Raman and LC/MS analyses indicate that TiO2 breaks the compound into piperazinyl and carboxylate groups which attach to the surface, whereas de-carboxylated and hydroxylated quinolone moieties remain in solution. The present study thus identifies the sorption mechanisms and breakdown products of OFL during dark reactions with TiO2, which is critically important for understanding the fate and transport of OFL as it enters the soil and aquatic environment.
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis