Merck
  • A comparative analysis of the distribution and composition of lipidic constituents and associated enzymes in pollen and stigma of sunflower.
A comparative analysis of the distribution and composition of lipidic constituents and associated enzymes in pollen and stigma of sunflower.
Sexual plant reproduction (2010-05-22)
Rashmi Shakya, Satish Chander Bhatla
ABSTRACT
Spatial distribution and compositional analyses of the lipidic constituents in pollen and stigma of sunflower (Helianthus annuus L. cv. Morden) were conducted using ultrastructural, histochemical, and biochemical analysis. Detection of secretions at the base of stigmatic papillae and neutral lipid accumulations on the surface of stigmatic papillae and between adjacent pseudopapillae demonstrates the semidry nature of stigma surface in sunflower. Pollen coat is richer in lipids (8%) than stigma (2.2%) on fresh weight basis. Nile Red-fluorescing neutral lipids are preferentially localized in the pollen coat. Neutral esters and triacylglycerols (TAGs) are the major lipidic constituents in pollen grains and stigma, respectively. Lignoceric acid (24:0) and cis-11-eicosenoic acid (20:1) are specifically expressed only in the pollen coat. Similar long-chain fatty acids have earlier been demonstrated to play a significant role during the initial signaling mechanism leading to hydration of pollen grains on the stigma surface. Lipase (EC 3.1.1.3) activity is expressed both in pollen grains and stigma. Stigma exhibits a better expression of acyl-ester hydrolase (EC 3.1.1.1) activity than that of observed in both the pollen fractions. Expression of two acyl-ester hydrolases (41 and 38 kDa) has been found to be specific to pollen coat. Specific expression of lignoceric acid (24:0) in pollen coat and localization of lipase in pollen and stigma have been discussed to assign possible roles that they might play during pollen-stigma interaction.
Sigma-Aldrich
Lignoceric acid, ≥99% (GC)