Merck
  • A protocol for analyzing the protein terminome of human cancer cell line culture supernatants.
A protocol for analyzing the protein terminome of human cancer cell line culture supernatants.
STAR protocols (2021-08-12)
Kazuya Tsumagari, Chih-Hsiang Chang, Yasushi Ishihama
ABSTRACT
Characterization of protein termini is essential for understanding how the proteome is generated through biological processes such as post-translational proteolytic events. Here, we introduce a practical protocol for terminomics to achieve simple and sensitive N- and C-terminal peptide enrichment. We apply it to the terminome analysis of culture supernatants of a human cancer cell line for the purpose of identifying ectodomain shedding substrate cleavage sites with 10 μg protein per sample. For complete details on the use and execution of this protocol, please refer to Tsumagari et al. (2021).
Sigma-Aldrich
LysargiNase, suitable for peptide or protein digestion insolution or in gel
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution