Merck
The aspartimide problem in Fmoc-based SPPS. Part I.
Journal of peptide science : an official publication of the European Peptide Society (2003-02-18)
M Mergler, F Dick, B Sax, P Weiler, T Vorherr
ABSTRACT
A variety of Asp beta-carboxy protecting groups, Hmb backbone protection and a range of Fmoc cleavage protocols have been employed in syntheses of the model hexapeptide H-VKDGYI-OH to investigate the aspartimide problem in more detail. The extent of formation of aspartimide and aspartimide-related by-products was determined by RP-HPLC. This study included three new Fmoc-Asp-OH derivatives: the beta-(4-pyridyl-diphenylmethyl) and beta-(9-phenyl-fluoren-9-yl) esters and also the orthoester Fmoc-beta-(4-methyl-2,6,7-trioxabicyclo[2.2.2]-oct-1-yl)-alanine. 3-Methylpent-3-yl protection of the Asp side chain resulted in significant improvements with respect to aspartimide formation. Complete suppression was achieved using the combination OtBu side chain protection and Hmb backbone protection for the preceding Gly residue.