Merck
  • Integrator enforces the fidelity of transcriptional termination at protein-coding genes.
Integrator enforces the fidelity of transcriptional termination at protein-coding genes.
Science advances (2021-11-04)
Lucas Ferreira Dasilva, Ezra Blumenthal, Felipe Beckedorff, Pradeep Reddy Cingaram, Helena Gomes Dos Santos, Raghu Ram Edupuganti, Anda Zhang, Sadat Dokaneheifard, Yuki Aoi, Jingyin Yue, Nina Kirstein, Mina Masoumeh Tayari, Ali Shilatifard, Ramin Shiekhattar
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Anti-INTS11 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution