Merck
  • Loss of miR-29a impairs decidualization of endometrial stromal cells by TET3 mediated demethylation of Col1A1 promoter.
Loss of miR-29a impairs decidualization of endometrial stromal cells by TET3 mediated demethylation of Col1A1 promoter.
iScience (2021-09-28)
Aixia Liu, Mengmeng Jin, Laidi Xie, Mengyu Jing, Ying Zhou, Minyue Tang, Tingting Lin, Dimin Wang
ABSTRACT
A conceptual framework for understanding abnormal endometrial decidualization, with considerable significance for the diagnosis and treatment of abnormal decidualization-related changes in non-receptive endometrium in implantation failure during early pregnancy is very important. Here, we found the expression levels of miR-29a in endometrial tissues were associated with the menstrual phases and pregnancy outcome. Inhibition of miR-29a led to decreased decidualization of endometrial stromal cells (ESCs) in vitro, whereas Tet methylcytosine dioxygenase 3 (TET3) and its potential demethylation target, the collagen type I alpha 1 chain (Col1A1), were restored. The binding capacity of TET3 to the Col1A1 promoter could be enhanced by the inhibition of miR-29a. Finally, deletion of TET3 rescued the inhibitory effect of the miR-29a antagomir on the proliferation of decidualized ESCs in vitro and embryo implantation in vivo. Thus, loss of miR-29a causes implantation failure because of the limitation of ESCs decidualization-related changes in non-receptive endometrium during early pregnancy.
Sigma-Aldrich
Anti-TET3, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-TET3 Antibody, from rabbit, purified by affinity chromatography