Merck
  • Urinary megalin deficiency implicates abnormal tubular endocytic function in Fanconi syndrome.
Urinary megalin deficiency implicates abnormal tubular endocytic function in Fanconi syndrome.
Journal of the American Society of Nephrology : JASN (2001-12-26)
Anthony G W Norden, Marta Lapsley, Takashi Igarashi, Catherine L Kelleher, Philip J Lee, Takeshi Matsuyama, Steven J Scheinman, Hiroshi Shiraga, David P Sundin, Rajesh V Thakker, Robert J Unwin, Pierre Verroust, Søren K Moestrup
ABSTRACT
Normal reabsorption of glomerular filtrate proteins probably requires recycling of the endocytic receptors megalin (gp330) and cubilin. Both receptors are located on the luminal surface of the renal proximal tubule epithelium. Whether abnormal amounts of receptor are present in the urine of patients with Dent's disease, Lowe's syndrome, or autosomal dominant idiopathic Fanconi syndrome was explored. They are all forms of the renal Fanconi syndrome and are associated with tubular proteinuria. Urine samples of equal creatinine contents were dialyzed, lyophilized, and subjected to electrophoresis on nonreducing sodium dodecyl sulfate-5% polyacrylamide gels. Proteins were blotted and probed with anti-megalin IgG, anti-cubilin IgG, or receptor-associated protein. Megalin and cubilin levels detected by immunochemiluminescence were measured as integrated pixels and expressed as percentages of the normal mean values. A striking deficiency of urinary megalin, compared with normal individuals (n = 42), was observed for eight of nine families with Dent's disease (n = 10) and for the two families with Lowe's syndrome (n = 3). The family with autosomal dominant idiopathic Fanconi syndrome (n = 2) exhibited megalin levels within the normal range. The measured levels of cubilin were normal for all patients. These results are consistent with defective recycling of megalin to the apical cell surface of the proximal tubules and thus decreased loss into urine in Dent's disease and Lowe's syndrome. This defect would interfere with the normal endocytic function of megalin, result in losses of potential ligands into the urine, and produce tubular proteinuria.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder