Merck
p53 induces ARTS to promote mitochondrial apoptosis.
Cell death & disease (2021-02-26)
Qian Hao, Jiaxiang Chen, Junming Liao, Yingdan Huang, Yu Gan, Sarit Larisch, Shelya X Zeng, Hua Lu, Xiang Zhou
ABSTRACT
Apoptosis related protein in TGF-β signaling pathway (ARTS) was originally discovered in cells undergoing apoptosis in response to TGF-β, but ARTS also acts downstream of many other apoptotic stimuli. ARTS induces apoptosis by antagonizing the anti-apoptotic proteins XIAP and Bcl-2. Here we identified the pro-apoptotic Sept4/ARTS gene as a p53-responsive target gene. Ectopic p53 and a variety of p53-inducing agents increased both mRNA and protein levels of ARTS, whereas ablation of p53 reduced ARTS expression in response to multiple stress conditions. Also, γ-irradiation induced p53-dependent ARTS expression in mice. Consistently, p53 binds to the responsive DNA element on the ARTS promoter and transcriptionally activated the promoter-driven expression of a luciferase reporter gene. Interestingly, ARTS binds to and sequesters p53 at mitochondria, enhancing the interaction of the latter with Bcl-XL. Ectopic ARTS markedly augments DNA damage stress- or Nutlin-3-triggered apoptosis, while ablation of ARTS preferentially impairs p53-induced apoptosis. Altogether, these findings demonstrate that ARTS collaborates with p53 in mitochondria-engaged apoptosis.
Sigma-Aldrich
Anti-ARTS antibody, Mouse monoclonal, clone ARTS51, purified from hybridoma cell culture
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)