Merck
  • HIV-1 uncoating by release of viral cDNA from capsid-like structures in the nucleus of infected cells.
HIV-1 uncoating by release of viral cDNA from capsid-like structures in the nucleus of infected cells.
eLife (2021-04-28)
Thorsten G Müller, Vojtech Zila, Kyra Peters, Sandra Schifferdecker, Mia Stanic, Bojana Lucic, Vibor Laketa, Marina Lusic, Barbara Müller, Hans-Georg Kräusslich
ABSTRACT
HIV-1 replication commences inside the cone-shaped viral capsid, but timing, localization, and mechanism of uncoating are under debate. We adapted a strategy to visualize individual reverse-transcribed HIV-1 cDNA molecules and their association with viral and cellular proteins using fluorescence and correlative-light-and-electron-microscopy (CLEM). We specifically detected HIV-1 cDNA inside nuclei, but not in the cytoplasm. Nuclear cDNA initially co-localized with a fluorescent integrase fusion (IN-FP) and the viral CA (capsid) protein, but cDNA-punctae separated from IN-FP/CA over time. This phenotype was conserved in primary HIV-1 target cells, with nuclear HIV-1 complexes exhibiting strong CA-signals in all cell types. CLEM revealed cone-shaped HIV-1 capsid-like structures and apparently broken capsid-remnants at the position of IN-FP signals and elongated chromatin-like structures in the position of viral cDNA punctae lacking IN-FP. Our data argue for nuclear uncoating by physical disruption rather than cooperative disassembly of the CA-lattice, followed by physical separation from the pre-integration complex.
Sigma-Aldrich
Anti-CPSF6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG - Atto 594 antibody produced in goat