Merck
  • Design, synthesis and evaluation of non-urea inhibitors of soluble epoxide hydrolase.
Design, synthesis and evaluation of non-urea inhibitors of soluble epoxide hydrolase.
Bioorganic & medicinal chemistry letters (2011-11-15)
Stevan Pecic, Shi-Xian Deng, Christophe Morisseau, Bruce D Hammock, Donald W Landry
ABSTRACT
Inhibition of soluble epoxide hydrolase (sEH) has been proposed as a new pharmaceutical approach for treating hypertension and vascular inflammation. The most potent sEH inhibitors reported in literature to date are urea derivatives. However, these compounds have limited pharmacokinetic profiles. We investigated non-urea amide derivatives as sEH inhibitors and identified a potent human sEH inhibitor 14-34 having potency comparable to urea-based inhibitors.
Sigma-Aldrich
N,N′-Dicyclohexylurea, 98%