Merck
  • High yield cyclotron production of a novel 133/135La theranostic pair for nuclear medicine.
High yield cyclotron production of a novel 133/135La theranostic pair for nuclear medicine.
Scientific reports (2020-12-19)
Bryce J B Nelson, John Wilson, Jan D Andersson, Frank Wuest
ABSTRACT
This study reports the high-yield production of a novel 133/135La theranostic pair at a 22 MeV proton beam energy as an attractive alternative to the recently introduced 132/135La pair, demonstrating over an order of magnitude production increase of 133/135La (231 ± 8 MBq 133La and 166 ± 5 MBq 135La at End of Bombardment (EOB)) compared to 11.9 MeV production of 132/135La (0.82 ± 0.06 MBq 132La and 19.0 ± 1.2 MBq 135La) for 500 µA·min irradiations. A new sealed solid cyclotron target is introduced, which is fast to assemble, easy to handle, storable, and contains reusable components. Radiolabeling with macrocyclic chelators DOTA and macropa achieved full incorporation, with respective apparent 133La molar activites of 33 ± 5 GBq/µmol and 30 ± 4 GBq/µmol. PET centers with access to a 22 MeV capable cyclotron could produce clinically-relevant doses of 133/135La, via natBa irradiation, as a standalone theranostic agent for PET imaging and Auger electron therapy. With lower positron energies and less energetic and abundant gamma rays than 68Ga, 44Sc and 132La, 133La appears to be an attractive radiometal candidate for PET applications requiring a higher scanning resolution, a relatively long isotopic half-life, ease of handling, and a low patient dose.
Sigma-Aldrich
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, ≥97.0% (CHN)
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis